missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ CD79a Polyclonal Antibody
GREENER_CHOICE

Produktkod. 30534221
Change view
Click to view available options
Kvantitet:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30534221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534221 Supplier Invitrogen™ Supplier No. PA5149277

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79a
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 0.2 mg/mL
Konjugera Unconjugated
Formulering PBS with trehalose and no preservative
Gen CD79A
Genaccessionsnr. P11911
Gene Alias B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein
Gensymboler CD79A
Värd art Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD79A, Leu29-Arg137.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 12518
Målarter Mouse
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Lyophilized
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.