missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BUV563, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
2575.00 SEK - 5745.00 SEK
Specifications
Antigen | CD8 |
---|---|
Clone | RPA-T8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 563 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16619037
|
BD Biosciences
612915 |
25 Tests |
2575.00 SEK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17267964
|
BD Biosciences
612914 |
100 Tests |
5745.00 SEK
100 tests |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Spécification
CD8 | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
RPA-T8 | |
Monoclonal | |
Mouse | |
Human, Baboon, Cynomolgus, Rhesus Monkey | |
Primary | |
CD8 |