missing translation for 'onlineSavingsMsg'
Learn More

CD8a Rat anti-Canine, APC, Clone: YCATE55.9, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5080-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568106

  • 986.00 SEK / 25 tests


Estimated Shipment
14-01-2021
Add to basket

Description

Description

CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications

Specifications

CD8a
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P33706
CD8, Leu-2, MAL, p32, CD8a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine
Flow Cytometry
YCATE55.9
APC
CD8A
Liquid
CD8A
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
403157
Videos
SDS
Documents

For Research Use Only.