missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD80 (Human) Recombinant Protein

Human CD80 full-length ORF ( AAH42665, 35 a.a. - 288 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000941-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132841

  • 6025.00 SEK / 25µg
Estimated Shipment: 08-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: VIHVTKEVKEVATLSCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDITNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPTPSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETELYAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDNLLPSWAITLISVNGIFVICCLTYCFAPRCRERRRNERLRRESVRPV
Specifications

Specifications

AAH42665
CD80 molecule
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD80
GST
941
Wheat germ expression system
25 μg
CD28LG, CD28LG1, LAB7
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers