missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Monoclonal Antibody (M38), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15781
Additional Details : Weight : 0.00280kg
Description
CD81 (TAPA-1, target of anti-proliferative antibody-1) is a member of the tetraspanin family, is expressed on virtually all nucleated cells, but above all on germinal center B cells. CD81 forms complexes with other tetraspanin proteins, integrins, coreceptors, MHC class I and II molecules, and influences adhesion, morphology, activation, proliferation and differentiation of B, T cells. In muscles, CD81 promotes cell fusion and myotube maintece. CD81 has been also identified as a receptor for the hepatitis C virus. Like members of the tetraspanin family that include CD9, CD37, CD53, CD63, and CD82, CD81 is a cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that is known to complex with integrins. CD81 appears to promote muscle cell fusion and support myotube maintece. CD81 associates with CD19, CD21, Leu 13, and integrins on cell membrane and acts as a receptor for the envelope protein E2 of chronic hepatitis C virus. Antibodies to CD81 have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. CD81 is also localized in the tumor-suppressor gene region and is a candidate gene for maligcies.Specifications
CD81 | |
Monoclonal | |
PE | |
CD81 | |
26 kDa cell surface protein TAPA-1, CD81 antigen, CD81 antigen, target of antiproliferative antibody 1, tetraspanin-28, tspan-28 | |
Mouse | |
Purified | |
RUO | |
975 | |
4° C | |
Liquid |
Flow Cytometry | |
M38 | |
PBS with 0.2% BSA and 15mM sodium azide | |
P60033 | |
CD81 | |
MOLT-4 (human T-ALL cell line) | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |