Learn More
CD85j (ILT2) Monoclonal Antibody (HP-F1), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience 48-5129-42
Weitere Details : Weight : 0.01000kg
Beschreibung
The monoclonal antibody HP-F1 recognizes CD85j, also known as ILT2, LILRB1, and LIR1. CD85j is a member of the ILT/LIR/MIR family. CD85j is a single transmembrane glycoprotein with a long cytoplasmic domain containing 4 ITIMs which signal through interactions with SHP-1. Expression is found on myeloid cells (monocytes and dendritic cells) and some lymphoid cells including, subsets of NK, T and B cells. Expression has been correlated with leukemias such as ALL and CLL. Expression on CD8+ cells correlates with effector cell function and plays an important role in viral infections, including HIV, Ebstein Barr and CMV. The ligands for CD85j are MHC Class I molecules such as HLA-G, A, F, B27, E and F. The monoclonal antibody HP-F1 has been shown to reduce the amount of CD16- dependent cytolytic activity of functional NK cells. Applications Reported: This HP-F1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HP-F1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5uL (0.25ug) per test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100uL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm).Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser
Description: The monoclonal antibody HP-F1 recognizes CD85j, also known as ILT2, LILRB1, and LIR1. CD85j is a member of the ILT (immunoglobulin-like transcript)/LIR (leukocyte Ig-like receptor)/MIR (monocyte Ig-like receptor) family. CD85j is a single transmembrane glycoprotein with a long cytoplasmic domain containing 4 ITIMs which signal through interactions with SHP-1. Expression is found on myeloid cells (monocytes and dendritic cells) and some lymphoid cells including, subsets of NK, T and B cells. Expression has been correlated with leukemias such as ALL and CLL. Expression on CD8+ cells correlates with effector cell function and plays an important role in viral infections, including HIV, Ebstein Barr and CMV. The ligands for CD85j are MHC Class I molecules such as HLA-G, A, F, B27, E and F. The monoclonal antibody HP-F1 has been shown to reduce the amount of CD16- dependent cytolytic activity of functional NK cells. Applications Reported: This HP-F1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HP-F1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet LaserSpezifikation
CD85j (ILT2) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q8NHL6 | |
LILRB1 | |
Affinity chromatography | |
RUO | |
10859 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HP-F1 | |
eFluor 450 | |
LILRB1 | |
CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |