missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Monoclonal Antibody (BU63), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD8601
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD86 Monoclonal antibody specifically detects CD86 in Human samples. It is validated for Flow Cytometry.Specifications
CD86, CD86 (B7-2) | |
Monoclonal | |
FITC | |
CD86 | |
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
Mouse | |
purified | |
ASR | |
942 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
BU63 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P42081 | |
CD86 | |
B-lymphoblastoid cell line ARH 77 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |