Läs mer
CD86 Rat anti-Mouse, PE, Clone: GL1, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech PE-65068-100UG
Beskrivning
The CD86 antibody from Proteintech is a rat monoclonal antibody generated with lps-activated cba/ca mouse spleen b cells of mouse CD86. This antibody recognizes mouse antigen. The CD86 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifikationer
| CD86 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| GL1 | |
| PE | |
| 100 μg | |
| 12524 | |
| IgG2a |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.