missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 alpha Monoclonal Antibody (53-6.7), APC-Cyanine7, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A15386
Additional Details : Weight : 0.00280kg
Description
CD8 alpha Monoclonal antibody specifically detects CD8 alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
CD8 alpha | |
Monoclonal | |
0.1 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide | |
P01731, P10300 | |
Cd8a, Cd8b1 | |
Mouse CD8a. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
53-6.7 | |
APC-Cyanine7 | |
Cd8b1 | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at | |
Rat | |
purified | |
RUO | |
12525, 12526 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |