missing translation for 'onlineSavingsMsg'
Learn More

CD8b Mouse anti-Human, Non-human primate, PE-Cyanine7, Clone: SIDI8BEE, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-5273-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568556

  • 2605.00 SEK / 100 tests


Estimated Shipment
02-12-2020
Add to basket

Description

Description

Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells.
Specifications

Specifications

CD8b
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P10966
Cd8b1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Non-human Primate
Flow Cytometry
SIDI8BEE
PE-Cyanine7
CD8B
Liquid
CD8B
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
926
Videos
SDS
Documents

For Research Use Only.