missing translation for 'onlineSavingsMsg'
Learn More

CD9 Mouse anti-Human, Rat, Clone: 4H7B9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60232-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877255

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. CD9 belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41 / CD61 (GPIIb / GPIIIa). CD9 is involved in cell motility, osteoclastogenesis, neurite outgrowth, myotube formation, and sperm-egg fusion, plays roles in cell attachment and proliferation and is necessary for association of heterologous MHC II molecules on the dendritic cell plasma membrane which is important for effective T cell stimulation. CD9 functions in many cellular processes including differentiation, adhesion, and signal transduction, and expression plays a critical role in the suppression of cancer cell motility and metastasis. CD9 is also considered as metastasis suppressor in solid tumors.
Specifications

Specifications

CD9
4H7B9
Unconjugated
CD9
Liquid
CD9
CD9 Fusion Protein Ag14529
Protein G
RUO
Antibody
Monoclonal
Human, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21926, P40241
5H9, 5H9 antigen, BA2, BTCC 1, CD9, CD9 antigen, CD9 molecule, DRAP 27, GIG2, Leukocyte antigen MIC3, MIC3, Motility related protein, MRP 1, P24, Tetraspanin 29, Tspan 29, TSPAN29
Mouse
IgG1
150 μL
-20°C
Primary
24936, 928
Videos
SDS
Documents
Certificates
Special Offers

Special Offers