missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 Monoclonal Antibody (G7), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14725
Additional Details : Weight : 0.01000kg
Description
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.Specifications
CD90 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P01831 | |
Thy1 | |
MST cell hybridoma C6/G8 from Mouse T cell hybridoma C6/G8 | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2c κ |
Flow Cytometry | |
G7 | |
FITC | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Rat | |
purified | |
RUO | |
21838 | |
4° C, store in dark | |
Liquid |