missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 Monoclonal Antibody (G7), FITC, Invitrogen™
Rat Monoclonal Antibody
1138.00 SEK - 5200.00 SEK
Specifications
Antigen | CD90 |
---|---|
Clone | G7 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13385564
|
Invitrogen
A14725 |
25 μg |
1138.00 SEK
25µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10757225
|
Invitrogen
MCD9001 |
500 μg |
5200.00 SEK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD90 Monoclonal antibody specifically detects CD90 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD90 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with <0.1% sodium azide | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Thy1 | |
IgG2c κ | |
purified | |
Antibody |
G7 | |
Flow Cytometry | |
FITC | |
Rat | |
Mouse | |
P01831 | |
21838 | |
MST cell hybridoma C6/G8 from Mouse T cell hybridoma C6/G8 | |
Primary | |
4° C, store in dark | |
Thy1 |