missing translation for 'onlineSavingsMsg'
Learn More

CD99 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23079-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822185

  • 1236.20 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell adhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the hematopoietic system to T, NK and myeloid cells.
Specifications

Specifications

CD99
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14209
12E7, CD99, CD99 antigen, CD99 molecule, E2 antigen, MIC2, MIC2X, MIC2Y, Protein MIC2, T cell surface glycoprotein E2
Rabbit
IgG
20 μL
-20°C
Primary
4267
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
CD99
Liquid
CD99
CD99 Fusion Protein Ag19461
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers