missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC123/C10orf7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28178-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid. The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2; MIM 248610), and a homodimeric dihydrolipoamide dehydrogenase (E3; MIM 238331). E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene (MIM 248611) encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.Specifications
CDC123/C10orf7 | |
Polyclonal | |
Unconjugated | |
CDC123 | |
C10orf7, CDC123, D123, FLJ13863, HT 1080, Protein D123, PZ32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8872 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75794 | |
CDC123 | |
CDC123/C10orf7 Fusion Protein Ag26828 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |