missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDH18 (Human) Recombinant Protein
Human CDH18 full-length ORF ( AAH31051, 1 a.a. - 790 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001016-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKITSTSCICPVLVCLCFVQRCYGTAHHSSIKVMRNQTKHIEGETEVHHRPKRGWVWNQFFVLEEHMGPDPQYVGKLHSNSDKGDGSVKYILTGEGAGTIFIIDDTTGDIHSTKSLDREQKTHYVLHAQAIDRRTNKPLEPESEFIIKVQDINDNAPKFTDGPYIVTVPEMSDMGTSVLQVTATDADDPTYGNSARVVYSILQGQPYFSVDPKTGVIRTALHNMDGEAREHYSVVIQAKDMAGQVGGLSGSTTVNITLTDVNDNTPRFPQKHYQLYVPESAQVGSAVGKIKANDADTGSNADMTYSIINGDGMGIFSISTDKETREGILSLKKPLNYEKKKSYTLNIEGANTHLDFRFSHLGPFKDATMLKIIVGDVDEPPLFSMPSYLMEVYENAKIGTVVGTVLAQDPDSINSLVRYFINYNVEDDRFFNIDANTGTIRTTKVLDREETPWYNITVTASEIDNPDLLSHVTVGIRVLDVNDNPPELAREYDIIVCENSKPGQVIHTISATDKDDFANGPRFNFFLDERLPVNPNFTLKDNEDNTASILTRRRRFSRTVQDVYYLPIMISDGGIPSLSSSSTLTIRVCACERDGRVRTCHAEAFLSSAGLSTGALIAILLCVLILLAIVVLFITLRRSKKEPLIISEEDVRENVVTYDDEGGGEEDTGAFDITALRNPSAAEELKYRRDIRPEVKLTPRHQTSSTLESIDVQEFIKQRLAEADLDPSVPPYDSLQTYAYEGQRSEAGSISSLDSATTQSDQDYHYLGDWGPEFKKLAELYGEIESERTT
Specifications
AAH31051 | |
cadherin 18, type 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDH18 | |
GST |
1016 | |
Wheat germ expression system | |
10 μg | |
CDH14, CDH14L, CDH24, EY-CADHERIN | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |