missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDH7 (Human) Recombinant Protein
Human CDH7 full-length ORF ( AAH36786, 1 a.a. - 630 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH36786 |
---|---|
Gene ID (Entrez) | 1005 |
Name | cadherin 7, type 2 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172981
|
Abnova™
H00001005-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182981
|
Abnova™
H00001005-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MKLGKVEFCHFLQLIALFLCFSGMSQAELSRSRSKPYFQSGRSRTKRSWVWNQFFVLEEYMGSDPLYVGKLHSDVDKGDGSIKYILSGEGASSIFIIDENTGDIHATKRLDREEQAYYTLRAQALDRLTNEPVEPESEFVIKIQDINDNEPKFLDGPYTAGVPEMSPVGTSVVQVTATDADDPTYGNSARVVYSILQGQPYFSVEPKTGVIKTALPNMDREAKDQYLLVIQAKDMVGQNGGLSGTTSVTVTLTDVNDNPPRFPRRSYQYNVPESLPVASVVARIKAADADIGANAEMEYKIVDGDGLGIFKISVDKETQEGIITIQKELDFEAKTSYTLRIEAANKDADPRFLSLGPFSDTTTVKIIVEDVDEPPVFSSPLYPMEVSEATQVGNIIGTVAAHDPDSSNSPVRYSIDRNTDLERYFNIDANSGVITTAKSLDRETNAIHNITVLAMESQNPSQVGRGYVAITILDINDNAPEFAMDYETTVCENAQPGQVIQKISAVDKDEPSNGHQFYFSLTTDATNNHNFSLKDNKDNTASILTRRNGFRRQEQSVYYLPIFIVDSGSPSLSSTSTLTIRVCDCDADGVAQTCNAEAYVLPAGLSTGALIAILACVLTLLGRYCFQGLL
Specifications
AAH36786 | |
cadherin 7, type 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
CDH7L1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1005 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDH7 | |
GST |