missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDH7 (Human) Recombinant Protein
Human CDH7 full-length ORF ( AAH36786, 1 a.a. - 630 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001005-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKLGKVEFCHFLQLIALFLCFSGMSQAELSRSRSKPYFQSGRSRTKRSWVWNQFFVLEEYMGSDPLYVGKLHSDVDKGDGSIKYILSGEGASSIFIIDENTGDIHATKRLDREEQAYYTLRAQALDRLTNEPVEPESEFVIKIQDINDNEPKFLDGPYTAGVPEMSPVGTSVVQVTATDADDPTYGNSARVVYSILQGQPYFSVEPKTGVIKTALPNMDREAKDQYLLVIQAKDMVGQNGGLSGTTSVTVTLTDVNDNPPRFPRRSYQYNVPESLPVASVVARIKAADADIGANAEMEYKIVDGDGLGIFKISVDKETQEGIITIQKELDFEAKTSYTLRIEAANKDADPRFLSLGPFSDTTTVKIIVEDVDEPPVFSSPLYPMEVSEATQVGNIIGTVAAHDPDSSNSPVRYSIDRNTDLERYFNIDANSGVITTAKSLDRETNAIHNITVLAMESQNPSQVGRGYVAITILDINDNAPEFAMDYETTVCENAQPGQVIQKISAVDKDEPSNGHQFYFSLTTDATNNHNFSLKDNKDNTASILTRRNGFRRQEQSVYYLPIFIVDSGSPSLSSTSTLTIRVCDCDADGVAQTCNAEAYVLPAGLSTGALIAILACVLTLLGRYCFQGLL
Specifications
AAH36786 | |
cadherin 7, type 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDH7 | |
GST |
1005 | |
Wheat germ expression system | |
25 μg | |
CDH7L1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |