missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CDK1/CDK2 Monoclonal Antibody (AN21.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA191380
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: HeLa nuclear extract.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
Specifikationer
| CDK1/CDK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P06493, P24941 | |
| CDK1, CDK2 | |
| Human cdk2. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| AN21.2 | |
| Unconjugated | |
| CDK1 | |
| A630093N05Rik; Cdc2; CDC28A; Cdc2a; cdc2-related protein kinase; Cdk1; CDK2; CDKN1; CDKN2; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control protein 2 homolog; cell division cycle 2 homolog A; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 2; cyclin dependent kinase 1; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 1; Cyclin-dependent kinase 2; p33 protein kinase; p33(CDK2); p34 protein kinase; p34>CDC2>; P34CDC2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1017, 983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction