missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5/p35 Recombinant Human Protein
CDK5/p35 Recombinant Human Protein
Brand: Invitrogen PV3000
Additional Details : Weight : 3.36000kg
Description
CDK5 is a cyclin-dependent kinase that plays a critical role in neuron development. The kinase is strongly activated when it complexes with neuron-specific activators, especially p35, but less strongly when it complexes with cyclins. This protein is coexpressed and copurified with p35.Specifications
NP_004926.1, NP_003876 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
36.7/37.8kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
CDK5/p35 | |
CDK5/p35 was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 5mM MgCl2, 100μM ATP, 100μM histone H1 peptide substrate (PKTPKKAKKL) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
CDK5/p35 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.2mg/mL | |
1020, 8851 | |
CDK5/p35 Recombinant Human Protein | |
–80°C | |
1 Each | |
CDK5/p35 Recombinant Human Protein | |
CDK and CHK | |
1115nmoles of phosphate transferred to histone H1 peptide substrate (PKTPKKAKKL) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.25μg/mL | |
His Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.