missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK6/Cyclin D1 Recombinant Human Protein
This protein is coexpressed and copurified with CyclinD1.
Brand: Invitrogen™ PR8422B
Additional Details : Weight : 0.01000kg
Description
CDK6 functions as a cell-cycle initiator protein.
Specifications
NP_001250 I NP_444284 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
CDK6/Cyclin D1, CCND1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
127.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CDK and CHK Kinase Family | |
Recombinant | |
Baculovirus |