Learn More
Invitrogen™ CDK9/Cyclin K Recombinant Human Protein
CDK9 is a serine/threonine kinase. This protein is coexpressed and copurified with CyclinK.
Brand: Invitrogen™ PR7510A
Additional Details : Weight : 0.01000kg
Description
CDK9 phosphorylates the C-terminal domain of the largest subunit of RNA polymerase II and is involved in cell cycle regulation.
Specifications
NP_001252 I NP_003849 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
CDK9/Cyclin K, TAK | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
91.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CDK and CHK Kinase Family | |
Recombinant | |
Baculovirus |