Läs mer
CDK9-IN-1, MedChemExpress
Produktkod. 16496116
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496116 100 mg
16485568 10 mg
16461737 50 mg
16458267 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496116 Leverantör Medchem Express Leverantörsnummer HY13231
MedChemExpress CDK9-IN-1 is a novel, selective CDK9 inhibitor for the treatment of HIV infection, with an IC50 of 39 nM for CDK9/CycT1, extracted from reference, compound 87.
CDK9-IN-1 is a novel, selective CDK9 inhibitor for the treatment of HIV infection, with an IC50 of 39 nM for CDK9/CycT1, extracted from reference, compound 87.
Specifikationer
Kemiskt namn eller material CDK9-IN-1
Fysisk form Solid
CAS 1415559-43-1
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C26H21N5O4S
För användning med (applikation) COVID-19-anti-virus
Löslighetsinformation DMSO : 100 mg/mL (200.18 mM; Need ultrasonic)
LEDER CC1=C(NS(C)(=O)=O)C=C(NC2=NC=NC(C3=CC=CC(N4C(C5=C(C=CC=C5)C4=O)=O)=C3)=C2)C=C1
Molekylvikt (g/mol) 499.54
Formel vikt 499.54
Procent renhet 98.52%