missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
CDKAL1 Polyclonal specifically detects CDKAL1 in Human samples. It is validated for Western Blot.
Specifikationer
Specifikationer
| Antigen | CDKAL1 |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Konjugera | DyLight 550 |
| Utspädning | Western Blot |
| Formulering | 50mM Sodium Borate |
| Gene Alias | CDK5 regulatory subunit associated protein 1-like 1, CDK5 regulatory subunit-associated protein 1-like 1, FLJ20342, FLJ46705, MGC75469 |
| Värd art | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human CDKAL1 fragment. |
| Reningsmetod | Antigen Affinity-purified |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?