Learn More
Abnova™ CDKN1A Recombinant Protein
Brand:  Abnova™ H00001026-P01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16133041
  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH00312
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
43.8
8
Glutathione Sepharose 4 Fast Flow
10 ug
MSEPAGDVRQNPCGSKACRRLFGPVDSEQLSRDCDALMAGCIQEARERWNFDFVTETPLEGDFAWERVRGLGLPKLYLPTGPRRGRDELGGGRRPGTSPALLQGTAEEDHVDLSLSCTLVPRSGEQAEGSPGGPGDSQGRKRRQTSMTDFYHSKRRLIFSKRKP
CAP20/CDKN1/CIP1/MDA-6/P21/SDI1/WAF1/p21CIP1
CDKN1A
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
1026
CDKN1A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDKN1A
Human
Recombinant
Solution