Läs mer
Tocris Bioscience™ CDN 1163
Beskrivning
CDN 1163 is a SERCA2 allosteric activator. Increases Ca2+-ATPase activity and Ca2+ uptake by ER microsomes from obese mice. Rescues HEK cells from ER stress-induced cell death. Reduces fasting glucose levels and adipose tissue weight, and increases energy expenditure in ob/ob mice. Also improves glucose tolerance and reverses hepatic steatosis in ob/ob mice. Neuroprotective and improves memory and cognition in a mouse Alzheimer's disease model. Orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-(1-Methylethoxy)-N-(2-methyl-8-quinolinyl)benzamide |
| CAS | 892711-75-0 |
| Kvantitet | 10 mg |
| Mål | Ca2+-ATPase Activators |
| Molekylformel | C20H20N2O2 |
| Formel vikt | 320.38 |
| Renhet | >98% |