missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDYL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CDYL |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897724
|
Proteintech
17763-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16887724
|
Proteintech
17763-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chromodomain Y is a primate-specific Y-chromosomal gene family expressed exclusively in the testis and implicated in infertility. Although the Y-linked genes are testis-specific, this autosomal gene is ubiquitously expressed. The Y-linked genes arose by retrotransposition of an mRNA from this gene, followed by amplification of the retroposed gene. Proteins encoded by this gene superfamily possess a chromodomain, a motif implicated in chromatin binding and gene suppression, and a catalytic domain believed to be involved in histone acetylation. Multiple proteins are encoded by transcript variants of this gene.Specifications
CDYL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6AYK9, Q9WTK2, Q9Y232 | |
12593, 361237, 9425 | |
CDYL Fusion Protein Ag12143 | |
Primary | |
-20°C | |
CDYL |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDY like, CDYL, CDYL1, chromodomain protein, Y like, Chromodomain Y like protein | |
Cdyl | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |