missing translation for 'onlineSavingsMsg'
Learn More

CDYL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17763-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887724

  • 3248.87 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Chromodomain Y is a primate-specific Y-chromosomal gene family expressed exclusively in the testis and implicated in infertility. Although the Y-linked genes are testis-specific, this autosomal gene is ubiquitously expressed. The Y-linked genes arose by retrotransposition of an mRNA from this gene, followed by amplification of the retroposed gene. Proteins encoded by this gene superfamily possess a chromodomain, a motif implicated in chromatin binding and gene suppression, and a catalytic domain believed to be involved in histone acetylation. Multiple proteins are encoded by transcript variants of this gene.
Specifications

Specifications

CDYL
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AYK9, Q9WTK2, Q9Y232
CDY like, CDYL, CDYL1, chromodomain protein, Y like, Chromodomain Y like protein
Rabbit
IgG
150 μL
-20°C
Primary
12593, 361237, 9425
Immunoprecipitation, Western Blot
Unconjugated
CDYL
Liquid
Cdyl
CDYL Fusion Protein Ag12143
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers