missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CEACAM3 Monoclonal Antibody (8D5F12)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548419
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CEACAM3 Monoclonal Antibody for ELISA, Flow, Western Blot
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifikationer
| CEACAM3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P40198, Q63111 | |
| CEACAM3 | |
| Purified recombinant fragment of human CEACAM3 (AA: 35-155) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 8D5F12 | |
| Unconjugated | |
| CEACAM3 | |
| carcinoembryonic antigen CGM1; Carcinoembryonic antigen gene family (CGM1); carcinoembryonic antigen gene family 4; carcinoembryonic antigen gene family member 1; Carcinoembryonic antigen gene family member 4 / (carcinoembryonic antigene-related protein); carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen-related cell adhesion molecule 3; CD66d; CD66d antigen; CEA; cea12; CEACAM3; ceacam3 {ECO:0000312; Ceacam5; Cear; Cgm1; Cgm4; CGM4AA; EG384557; nonspecific cross-reacting antigen; pregnancy-specific glycoprotein 24; Psg24; RATCEAA; RATCGM4AA; RGD:2333}; W264; W282 | |
| Mouse | |
| Protein G | |
| RUO | |
| 1084, 24926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering