Läs mer
Tocris Bioscience™ Ceapin A7
Beskrivning
Ceapin A7 is a selective inhibitor of ATF6α (IC50=0.59 μM). The compound selectively inhibits ATF6α over ATF6β and other branches of the unfolded protein response (UPR). Ceapin A7 inhibits ER stress-induced upregulation of BiP in vitro. Ceapin A7 inhibits cleavage and functional activation of endogenous ATF6 in response to ER stress in vitro.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[1-[[2,4-bis(Trifluoromethyl)phenyl]methyl]-1 H-pyrazol-4-yl]-5-(2-furanyl)-3-isoxazolecarboxamide |
| CAS | 2323027-38-7 |
| Kvantitet | 50 mg |
| Mål | Other ER Stress/UPR Activators |
| Molekylformel | C20H12F6N4O3 |
| Renhet | 0.98 |