Läs mer
Cefacetrile Sodium, TRC
Produktkod. 30136418
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136418 50 mg
17954055 25 mg
17971825 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136418 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC23150550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23239-41-0
Molekylformel C13 H12 N3 O6 S . Na
Molekylvikt (g/mol) 361.31
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[(cyanoacetyl)amino]-8-oxo-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[(cyanoacetyl)amino]-8-oxo-, monosodium salt, (6R-trans)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-cyanoacetamido)-3-(hydroxymethyl)-8-oxo-, acetate (ester), monosodium salt (8CI), 7-Cyanoacetylaminocephalosporanic acid sodium salt, Ba 36278, CIBA 36278Ba, Cefacetrile sodium, Celospor, Celtol, Cephacetrile sodium, Cephacetrile-Na, Masticef, Sodium 3-(acetoxymethyl)-7-(2-cyanoacetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Sodium 7-(2-cyanoacetamido)cephalosporanic acid, Sodium 7-cyanacetamidocephalosporanate, Sodium cefacetril, Sodium cefacetrile, Sodium cephacetrile, Vetimast
IUPAC-namn sodium;(6R,7R)-3-(acetyloxymethyl)-7-[(2-cyanoacetyl)amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
LEDER [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC#N)C2=O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material Cefacetrile Sodium
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C13H13N3O6S.Na/c1-6(17)22-4-7-5-23-12-9(15-8(18)2-3-14)11(19)16(12)10(7)13(20)21;/h9,12H,2,4-5H2,1H3,(H,15,18)(H,20,21);/q;+1/p-1/t9-,12-;/m1./s1
Formel vikt 361.0345
Procent renhet >95%