Läs mer
Cefaclor Monohydrate, TRC
Produktkod. 30137453
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137453 250 mg
30132640 1 g
17933315 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137453 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC235250250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70356-03-5
Molekylformel C15 H14 Cl N3 O4 S . H2 O
Molekylvikt (g/mol) 385.82
Synonym Cefaclor Monohydrate, (6R,7R)-7-[[(2R)-Aminophenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monohydrate, 7-(D-2-amino-2-phenylacetamido)-3-chloro-3-cephem-4-carboxylic acid monohydrate
IUPAC-namn (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
LEDER O.N[C@@H](C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)Cl)c3ccccc3
Specifikationer
Kemiskt namn eller material Cefaclor Monohydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C15H14ClN3O4S.H2O/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7;/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23);1H2/t9-,10-,14-;/m1./s1
Formel vikt 385.0499
Procent renhet >95%