Läs mer
Cefamandole nafate, MedChemExpress
Produktkod. 16472256
Encompass_Preferred
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16472256 500 mg
16428656 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16472256 Leverantör Medchem Express Leverantörsnummer HYB1166
MedChemExpress Cefamandole nafate (Cefamandole formate sodium) is a second-generation broad-spectrum cephalosporin antibiotic.
Cefamandole nafate (Cefamandole formate sodium) is a second-generation broad-spectrum cephalosporin antibiotic.
Specifikationer
Kemiskt namn eller material Cefamandole nafate
Fysisk form Solid
CAS 42540-40-9
Färg Off-White
Kvantitet 500 mg
Anteckningar om renhetsgrad Research
Molekylformel C19H17N6NaO6S2
För användning med (applikation) COVID-19-immunoregulation
Synonym Cefamandole formate sodium
Löslighetsinformation DMSO : 100 mg/mL (195.13 mM; Need ultrasonic) ∣H2O : 100 mg/mL (195.13 mM; Need ultrasonic)
LEDER O=C(C(N12)=C(CSC3=NN=NN3C)CS[C@]2([H])[C@H](NC([C@H](OC=O)C4=CC=CC=C4)=O)C1=O)O[Na]
Molekylvikt (g/mol) 512.49
Formel vikt 512.49
Procent renhet 97.0%