missing translation for 'onlineSavingsMsg'
Läs mer

Cefdinir Sulfoxide, TRC

Produktkod. 30364107
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364107 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30364107 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC24269010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Cefdinir Sulfoxide
CAS 934986-48-8
Molekylformel C14 H13 N5 O6 S2
Kvantitet 10 mg
Synonym (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-(hydroxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid 5-oxide
InChI formel InChI=1S/C14H13N5O6S2/c1-2-5-4-27(25)12-8(11(21)19(12)9(5)13(22)23)17-10(20)7(18-24)6-3-26-14(15)16-6/h2-3,8,12,24H,1,4H2,(H2,15,16)(H,17,20)(H,22,23)/b18-7-/t8-,12-,27?/m1/s1
LEDER Nc1nc(cs1)\C(=N\O)\C(=O)N[C@H]2[C@@H]3N(C2=O)C(=C(CS3=O)C=C)C(=O)O
IUPAC-namn (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-5,8-dioxo-5λ^{4}-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molekylvikt (g/mol) 411.41
Formel vikt 411.0307
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.