Läs mer
Cefetamet Pivoxil, TRC
Produktkod. 30359588
Change view
Kvantitet:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359588 1 g
30364822 10 g
30348949 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359588 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2427801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65243-33-6
Molekylformel C20 H25 N5 O7 S2
Molekylvikt (g/mol) 511.57
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-methyl-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-methyl-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-methyl-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, (6R,7R)- (9CI), (2,2-Dimethyl-1-oxopropoxy)methyl (6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefetamet pivoxil, Ceftamet pivoxil
IUPAC-namn 2,2-dimethylpropanoyloxymethyl (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
LEDER CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)C)\c3csc(N)n3
Specifikationer
Kemiskt namn eller material Cefetamet Pivoxil
Kvantitet 1 g
InChI formel InChI=1S/C20H25N5O7S2/c1-9-6-33-16-12(23-14(26)11(24-30-5)10-7-34-19(21)22-10)15(27)25(16)13(9)17(28)31-8-32-18(29)20(2,3)4/h7,12,16H,6,8H2,1-5H3,(H2,21,22)(H,23,26)/b24-11-/t12-,16-/m1/s1
Formel vikt 511.1195