Läs mer
Cefmenoxime Hydrochloride, TRC
Produktkod. 30361228
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30361228 250 mg
1 options
Produktkod. 30361228 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC242835250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cefmenoxime HCl
CAS 75738-58-8
Molekylformel 2 C16 H17 N9 O5 S3 . Cl H
Kvantitet 250 mg
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, hydrochloride (2:1), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, hydrochloride (2:1), [6R-[6α,7β(Z)]]-, Abbott 50192, Bestcall, Bestron, Cefmax, Cefmenoxime hemihydrochloride, Cefmenoxime hydrochloride, Cemix, Tacef
InChI formel InChI=1S/2C16H17N9O5S3.ClH/c2*1-24-16(20-22-23-24)33-4-6-3-31-13-9(12(27)25(13)10(6)14(28)29)19-11(26)8(21-30-2)7-5-32-15(17)18-7;/h2*5,9,13H,3-4H2,1-2H3,(H2,17,18)(H,19,26)(H,28,29);1H/b2*21-8-;/t2*9-,13-;/m11./s1
LEDER Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CSc3nnnn3C)\c4csc(N)n4.CO\N=C(/C(=O)N[C@H]5[C@H]6SCC(=C(N6C5=O)C(=O)O)CSc7nnnn7C)\c8csc(N)n8
IUPAC-namn (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 1059.58
Formel vikt 1058.0796