Läs mer
Cefoperazone Sodium Salt, TRC
Produktkod. 30133336
Change view
Kvantitet:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133336 25 g
17950355 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133336 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC24290025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 62893-20-3
Molekylformel C25 H26 N9 O8 S2 . Na
Molekylvikt (g/mol) 667.65
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-[[(4-ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino](4-hydroxyphenyl)acetyl]amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[[[(4-ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino](4-hydroxyphenyl)acetyl]amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, monosodium salt, [6R-[6α,7β(R*)]]-, Bioperazone, CP 52640-2, Cefazone, Cefobine, Cefobis, Cefogram, Cefomycin, Cefoneg, Cefoperazone sodium, Cefoperazone sodium salt, Cefosint, Dardum, Farecef, Kefazon, Myticef, Novobiocyl, Pathozone, Peracef, Perocef, Sodium cefoperazone, T 1551, Tomabef
IUPAC-namn sodium;(6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
LEDER [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)[O-])CSc4nnnn4C)c5ccc(O)cc5)C(=O)C1=O
Specifikationer
Kemiskt namn eller material Cefoperazone sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 25 g
InChI formel InChI=1S/C25H27N9O8S2.Na/c1-3-32-8-9-33(21(39)20(32)38)24(42)27-15(12-4-6-14(35)7-5-12)18(36)26-16-19(37)34-17(23(40)41)13(10-43-22(16)34)11-44-25-28-29-30-31(25)2;/h4-7,15-16,22,35H,3,8-11H2,1-2H3,(H,26,36)(H,27,42)(H,40,41);/q;+1/p-1/t15-,16-,22-;/m1./s1
Formel vikt 667.1243
Procent renhet >95%