missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefotaxime Sodium Salt, ≥95%, MP Biomedicals™
Click to view available options
Quantity:
0.1 g
500 mg
Specifications
Type | Cefotaxime Sodium Salt |
Physical Form | Powder |
Assay Percent Range | ≥95% |
IUPAC Name | sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Formula Weight | 477.5 |
Color | White |
Quantity | 500 mg |
Chemical Name or Material | Cefotaxime Sodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'