Läs mer
Cefotiam hydrochloride, MedChemExpress
Produktkod. 16498588
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16498588 10 mg
16414467 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16498588 Leverantör Medchem Express Leverantörsnummer HYB0734A
MedChemExpress Cefotiam (SCE-963) hydrochloride is a parenteral cephalosporin antibiotic. Cefotiam hydrochloride has broad-spectrum activity against Gram-positive and Gram-negative bacteria.
Cefotiam (SCE-963) hydrochloride is a parenteral cephalosporin antibiotic. Cefotiam hydrochloride has broad-spectrum activity against Gram-positive and Gram-negative bacteria.
Specifikationer
Kemiskt namn eller material Cefotiam hydrochloride
Fysisk form Solid
CAS 66309-69-1
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H25Cl2N9O4S3
För användning med (applikation) Neuroscience-Neuromodulation
Synonym SCE-963 hydrochloride
Löslighetsinformation DMSO : 100 mg/mL (167.07 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (55.68 mM; Need ultrasonic)
LEDER O=C(C(N12)=C(CSC3=NN=NN3CCN(C)C)CS[C@]2([H])[C@H](NC(CC4=CSC(N)=N4)=O)C1=O)O.[H]Cl.[H]Cl
Molekylvikt (g/mol) 598.55
Formel vikt 598.55
Procent renhet 98.0%