Läs mer
Cefoxitin sodium, MedChemExpress
Produktkod. 16489875
Encompass_Preferred
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
16489875 250 mg
1 options
Produktkod. 16489875 Leverantör Medchem Express Leverantörsnummer HYB1117
MedChemExpress Cefoxitin sodium (MK-306) is a cephamycin antibiotic, often grouped with the second generation cephalosporins, acts by interfering with cell wall synthesis, its activity spectrum includes a broad range of gram-negative and gram-positive bacteria.
Cefoxitin sodium (MK-306) is a cephamycin antibiotic, often grouped with the second generation cephalosporins, acts by interfering with cell wall synthesis, its activity spectrum includes a broad range of gram-negative and gram-positive bacteria.
Specifikationer
Kemiskt namn eller material Cefoxitin sodium
Fysisk form Solid
CAS 33564-30-6
Färg White
Kvantitet 250 mg
Anteckningar om renhetsgrad Research
Molekylformel C16H16N3NaO7S2
För användning med (applikation) COVID-19-immunoregulation
Synonym MK-306
Löslighetsinformation DMSO : ≥ 100 mg/mL (222.50 mM) ∣H2O : ≥ 100 mg/mL (222.50 mM)
LEDER O=C(C(N12)=C(COC(N)=O)CS[C@]2([H])[C@@](NC(CC3=CC=CS3)=O)(OC)C1=O)O[Na]
Molekylvikt (g/mol) 449.43
Formel vikt 449.43
Procent renhet 99.43%