Läs mer
Cefpirome Sulfate, TRC
Produktkod. 30133653
Change view
Kvantitet:
100 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133653 1 g
30137239 2 g
17921215 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2435001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98753-19-6
Molekylformel C22 H22 N6 O5 S2 . H2 O4 S
Molekylvikt (g/mol) 612.66
Synonym Broact, Cefpirome sulfate, HR 810 sulfate, Keiten, 5H-Cyclopenta[b]pyridinium, 1-[[(6R,7R)-7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-6,7-dihydro-, sulfate (1:1) (9CI, ACI)
IUPAC-namn (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;sulfuric acid
LEDER CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])C[n+]3cccc4CCCc34)\c5csc(N)n5.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Cefpirome sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C22H22N6O5S2.H2O4S/c1-33-26-15(13-10-35-22(23)24-13)18(29)25-16-19(30)28-17(21(31)32)12(9-34-20(16)28)8-27-7-3-5-11-4-2-6-14(11)27;1-5(2,3)4/h3,5,7,10,16,20H,2,4,6,8-9H2,1H3,(H3-,23,24,25,29,31,32);(H2,1,2,3,4)/b26-15-;/t16-,20-;/m1./s1
Formel vikt 612.0767
Procent renhet >95%