Learn More
Cefpodoxime, 98%
Product Code. 15437499
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
15437499 1 g 1g
1 options
Product Code. 15437499 Supplier Thermo Scientific Alfa Aesar Supplier No. J66225.03
 Restricted Item
I lager
Add to basket
CAS: 80210-62-4 | C15H17N5O6S2 | 427.45 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Type Cefpodoxime
Physical Form Powder
CAS 80210-62-4
Melting Point 200°C to 202°C
Color Brown
Quantity 1 g
Sensitivity Light sensitive
Molecular Formula C15H17N5O6S2
MDL Number MFCD00864906
Synonym (6R,7R)-7-{2-(2-Amino-4-thiazolyl)-2-[(Z)-methoxyimino]acetamido}-3-(methoxymethyl)-3-cephem-4-carboxylate
Solubility Information Soluble in DMSO
InChI Key WYUSVOMTXWRGEK-LKXIZDLNSA-N
SMILES COCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)O
IUPAC Name (6S,7S)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular Weight (g/mol) 427.45
PubChem CID 124389585
Formula Weight 427.45
Percent Purity 98%
Chemical Name or Material Cefpodoxime