Learn More
Melford Cefsulodin
Product Code. 30018173
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018173 5 g 5g
30018171 100 mg 100mg
30018172 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30018173 Supplier Melford Supplier No. C520005.0
 Restricted Item
Beräknad leverans: 26-11-2026
Cefsulodin Sodium Salt is a cephalosporin antibiotic. Cefsulodin inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).
This disruption interferes with the final stages of peptidoglycan synthesis in the bacterial cell wall, leading to structural instability and eventual cell lysis.
Effective against a range of Gram-negative bacteria, it is particularly useful in the isolation and selection of these bacteria in microbiological and research settings. Commonly used in culture media for the growth and identification of specific bacterial strains.
Additionally, it is often employed as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion allows for the specific growth of Gram-negative bacteria lacking resistance to cefsulodin.
Specifications
Chemical Name or Material Cefsulodin
CAS 52152-93-9
Quantity 5 g
Molecular Formula C22H19N4NaO8S2