Läs mer
Ceftazidime Methyl Ester (Contains ~10% triethylammonium iodide), TRC
Produktkod. 30365745
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30365745 5 mg
1 options
Produktkod. 30365745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2441205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ceftazidime Methyl Ester
CAS 1354396-23-8
Molekylformel C23 H24 N6 O7 S2
Kvantitet 5 mg
Synonym Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(2-methoxy-1,1-dimethyl-2-oxoethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt, Ceftazidime pentahydrate Imp.H
InChI formel InChI=1S/C23H24N6O7S2/c1-23(2,21(34)35-3)36-27-14(13-11-38-22(24)25-13)17(30)26-15-18(31)29-16(20(32)33)12(10-37-19(15)29)9-28-7-5-4-6-8-28/h4-8,11,15,19H,9-10H2,1-3H3,(H3-,24,25,26,30,32,33)/b27-14-/t15-,19-/m1/s1
LEDER COC(=O)C(C)(C)O\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])C[n+]3ccccc3)\c4csc(N)n4
IUPAC-namn (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(1-methoxy-2-methyl-1-oxopropan-2-yl)oxyiminoacetyl]amino]-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molekylvikt (g/mol) 560.6
Formel vikt 560.1148