Läs mer
Cefteram Pivoxil, TRC
Produktkod. 30366638
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366638 5 mg
30360899 100 mg
30367278 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366638 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2443005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82547-81-7
Molekylformel C22 H27 N9 O7 S2
Molekylvikt (g/mol) 593.64
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, (6R,7R)- (9CI), Antibiotic T 2588, Cefteram pivoxil, Ro 19-5248, T 2588, Tomiron, Tomiron 100
IUPAC-namn 2,2-dimethylpropanoyloxymethyl (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(5-methyltetrazol-2-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
LEDER CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)Cn3nnc(C)n3)\c4csc(N)n4
Specifikationer
Kemiskt namn eller material Cefteram Pivoxil
Kvantitet 5 mg
InChI formel InChI=1S/C22H27N9O7S2/c1-10-26-29-30(27-10)6-11-7-39-18-14(25-16(32)13(28-36-5)12-8-40-21(23)24-12)17(33)31(18)15(11)19(34)37-9-38-20(35)22(2,3)4/h8,14,18H,6-7,9H2,1-5H3,(H2,23,24)(H,25,32)/b28-13-/t14-,18-/m1/s1
Formel vikt 593.1475