Läs mer
Cefuracetime (E-isomer) Sodium Salt, TRC
Produktkod. 30352383
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30352383 100 mg
1 options
Produktkod. 30352383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC243940100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-Cefuracetime sodium salt
CAS 97170-20-2
Molekylformel C17 H16 N3 O8 S . Na
Kvantitet 100 mg
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, monosodium salt, [6R-[6α,7β(E)]]- (9CI), Cefuracetime sodium (E-isomer), (E)-Cefuracetime sodium salt, (6R,7R)-7-[[(E)-(Furan-2-yl)(methoxyimino)acetyl]amino]-3-(hydroxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, (6R,7R)-3-[(Acetyloxy)methyl]-7-[[(2E)-2-furanyl(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt
InChI formel InChI=1S/C17H17N3O8S.Na/c1-8(21)28-6-9-7-29-16-12(15(23)20(16)13(9)17(24)25)18-14(22)11(19-26-2)10-4-3-5-27-10;/h3-5,12,16H,6-7H2,1-2H3,(H,18,22)(H,24,25);/q;+1/p-1/b19-11+;/t12-,16-;/m1./s1
LEDER [Na+].CO\N=C(\C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])COC(=O)C)/c3occc3
IUPAC-namn sodium;(6R,7R)-3-(acetyloxymethyl)-7-[[(2E)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molekylvikt (g/mol) 445.38
Formel vikt 445.0556