Läs mer
Cefuroxime Axetil Impurity C (85%), TRC
Produktkod. 30367382
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30367382 1 mg
1 options
Produktkod. 30367382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2480801MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cefuroxime Axetil Impurity C
CAS 76598-06-6
Molekylformel C18 H15 Cl3 N4 O9 S
Kvantitet 1 mg
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-3-[[[[(trichloroacetyl)amino]carbonyl]oxy]methyl]-, [6R-[6α,7β(Z)]]- (9CI), [6R-[6α,7β(Z)]]-7-[[2-Furanyl(methoxyimino)acetyl]amino]-8-oxo-3-[[[[(trichloroacetyl)amino]carbonyl]oxy]methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, Cefuroxime Axetil EP Impurity C(Cefuroxime Sodium EP D), (6R,7R)-7-[((Z)-(Furan-2-yl)(methoxyimino)acetyl)amino]-8-oxo-3-[([(trichloroacetyl)carbamoyl]oxy)methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic AcidCefuroxime Axetil Impurity C
InChI formel InChI=1S/C18H15Cl3N4O9S/c1-32-24-9(8-3-2-4-33-8)12(26)22-10-13(27)25-11(15(28)29)7(6-35-14(10)25)5-34-17(31)23-16(30)18(19,20)21/h2-4,10,14H,5-6H2,1H3,(H,22,26)(H,28,29)(H,23,30,31)/b24-9-/t10-,14-/m1/s1
LEDER CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)COC(=O)NC(=O)C(Cl)(Cl)Cl)\c3occc3
IUPAC-namn (6R,7R)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-3-[(2,2,2-trichloroacetyl)carbamoyloxymethyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molekylvikt (g/mol) 569.7571
Formel vikt 567.9625