Läs mer
Celiprolol EP Impurity A, Mikromol™
Produktkod. 30051028
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051028 25 mg
30049977 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051028 Leverantör Mikromol Leverantörsnummer MM0128.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Celiprolol EP Impurity A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-[5-Amino-2-[(2RS)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]ethanone
CAS 56980-94-0
Analyt- eller komponentnamn 1-[5-Amino-2-[(2RS)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]ethanone
Frakt skick Ice pack (-20°C)
Molekylformel C15 H24 N2 O3
Certifikat och efterlevnad ISO 9001
Synonym Celiprolol Hydrochloride Imp. A (EP), Celiprolol Imp. A (EP), 1-[5-Amino-2-[(2RS)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]ethanone, Celiprolol Hydrochloride Impurity A, Celiprolol Impurity A
InChI formel InChI=1S/C15H24N2O3/c1-10(18)13-7-11(16)5-6-14(13)20-9-12(19)8-17-15(2,3)4/h5-7,12,17,19H,8-9,16H2,1-4H3
LEDER CC(=O)c1cc(N)ccc1OCC(O)CNC(C)(C)C
IUPAC-namn 1-[5-amino-2-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]ethanone
Molekylvikt (g/mol) 280.36
Kvantitet 25 mg
Formel vikt 280.1787 g/mol