Learn More
CRABP2 Antibody, PE, R&D Systems™
Product Code. 18601526 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. Unit Size
18601526 0.1mL
1 options
Product Code. 18601526 Supplier Novus Biologicals Supplier No. NBP299740PE
 Restricted Item
Beräknad leverans: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
CRABP2 Polyclonal specifically detects CRABP2 in Human samples. It is validated for ELISA/Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Applications ELISA, Immunoprecipitation, Western Blot
Conjugate PE
Formulation PBS
Immunogen Produced in rabbits immunized with purified, recombinant Human CRABP2 (Accession#: NP_001869.1; Pro2-Glu 138)
Content And Storage Store at 4C in the dark.
Product Type Antibody
Product Line Novus