missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Cell Culture Flasks with Vented Filter Cap, Tissue Culture Treated

Preferred vessel for long term cultivation or large-scale expansion of cells, providing protection against contamination. Eppendorf™ Cell Culture Flasks with Vented Filter Cap, Tissue Culture Treated are sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free and non-cytotoxic.

Brand:  Eppendorf™ 0030712129

Additional Details : Weight : 6.66000kg

 View more versions of this product

Product Code. 15660527

  • 1310.00 SEK / Pack of 48


Estimated Shipment
02-09-2020
Add to basket

Description

Description

  • Features ConvexAcces™ geometry for facilitated access to growth area and easier and safe cell treatment
  • Optimized gas exchange and protection against contamination through high efficiency air filter technology
  • Anti-rolling cap with corrugation facilitates cultivation steps when cap needs to be set aside
  • 100% in-line control for leakage-free flasks and maximum safety
  • Direct surface identification on flask and cap
  • Available in three sizes: T–25, T–75 and T–175 with TC treated surfaces
Specifications

Specifications

NotAutoclavable
SAL10-6
Transparent
30 mL
  • Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene
  • Performance of cell-based assays and microscopic analysis
T-175 Cell culture flask
-86°C to +60°C
120.0 g
Ventedfiltercap
35.2 mm
179.5 cm2
Polystyrene
48
Tissue Culture Treated
20.0 to 30.0 mL
Videos
Documents